Assay Design
Gene Transcripts
- Gene Symbol
- sra-27
- Entrez Gene ID
-
184633
- Gene Name
- Serpentine receptor class alpha-27
- Gene Aliases
-
CELE_F18C5.1
- Chromosome Location
-
Chr.II: 6569007 - 6570701 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062927.3 |
NP_495328.2 |
6 - 7 |
882 |
165 |
View Full Product Details