Assay Design



Gene Transcripts

Gene Symbol
sra-29
Entrez Gene ID
24104447
Gene Name
Serpentine receptor class alpha-29
Gene Aliases
CELE_F18C5.8
Chromosome Location
Chr.II: 6574378 - 6575747 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306576.1 NP_001293505.1 1 - 2 240 159

View Full Product Details