Assay Design
Gene Transcripts
- Gene Symbol
- rpy-1
- Entrez Gene ID
-
174105
- Gene Name
- 43 kDa receptor-associated protein of the synapse homolog
- Gene Aliases
-
CELE_C18H9.7
- Chromosome Location
-
Chr.II: 6702213 - 6705657 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383869.1 |
|
3 - 4 |
274 |
82 |
View Full Product Details