Assay Design



Gene Transcripts

Gene Symbol
agr-1
Entrez Gene ID
3565243
Gene Name
AGRin (synaptic protein) homolog
Gene Aliases
CELE_F41G3.12
Chromosome Location
Chr.II: 6760674 - 6775176 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026981.6 NP_001022152.3 22 - 23 3548 77
GenBank mRNA AM773423.1 - 22 - 23 3548 77

View Full Product Details