Assay Design



Gene Transcripts

Gene Symbol
snt-1
Entrez Gene ID
174120
Gene Name
protein-coding
Gene Aliases
CELE_F31E8.2
Chromosome Location
Chr.II: 6810963 - 6816463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026958.5 NP_001022129.1 3 - 4 406 125
NM_062993.8 NP_495394.3 3 - 4 401 125
GenBank mRNA L15302.1 - 3 - 4 428 125

View Full Product Details