Assay Design
Gene Transcripts
- Gene Symbol
- aff-1
- Entrez Gene ID
-
174123
- Gene Name
- Cell fusion protein aff-1
- Gene Aliases
-
CELE_C44B7.3
- Chromosome Location
-
Chr.II: 6860582 - 6863752 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063001.6 |
NP_495402.3 |
9 - 10 |
1321 |
109 |
GenBank mRNA |
EF205023.1 |
- |
9 - 10 |
1321 |
109 |
View Full Product Details