Assay Design
Gene Transcripts
- Gene Symbol
- eif-2Bgamma
- Entrez Gene ID
-
174138
- Gene Name
- Probable translation initiation factor eIF-2B subunit gamma
- Gene Aliases
-
CELE_C15F1.4
- Chromosome Location
-
Chr.II: 6965797 - 6967648 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063027.8 |
NP_495428.1 |
2 - 3 |
324 |
87 |
View Full Product Details