Assay Design
Gene Transcripts
- Gene Symbol
- cutl-5
- Entrez Gene ID
-
184302
- Gene Name
- ZP domain-containing protein
- Gene Aliases
-
CELE_F10E7.10
- Chromosome Location
-
Chr.II: 7113683 - 7115906 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063071.2 |
NP_495472.1 |
1 - 2 |
150 |
124 |
View Full Product Details