Assay Design
Gene Transcripts
- Gene Symbol
- gld-3
- Entrez Gene ID
-
174174
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T07F8.3
- Chromosome Location
-
Chr.II: 7132643 - 7137522 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063078.4 |
NP_495479.2 |
16 - 17 |
2040 |
63 |
View Full Product Details