Assay Design



Gene Transcripts

Gene Symbol
acdh-12
Entrez Gene ID
174180
Gene Name
Acyl CoA DeHydrogenase
Gene Aliases
CELE_E04F6.5
Chromosome Location
Chr.II: 7192429 - 7195641 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026891.6 NP_001022062.1 3 - 4 314 91
NR_151558.1 2 - 3 273 91

View Full Product Details