Assay Design



Gene Transcripts

Gene Symbol
clh-3
Entrez Gene ID
174187
Gene Name
protein-coding
Gene Aliases
CELE_E04F6.11
Chromosome Location
Chr.II: 7217943 - 7225914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026889.5 NP_001022060.1 4 - 5 359 95
NM_001313600.3 NP_001300529.1 3 - 3 247 95
NM_001313601.3 NP_001300530.1 3 - 3 220 95
NM_001313602.3 NP_001300531.1 2 - 3 130 95
NM_001313604.3 NP_001300533.1 3 - 3 220 95
NM_001313605.3 NP_001300534.1 2 - 3 130 95
NM_001313606.3 NP_001300535.1 3 - 4 247 95
NR_132585.1 3 - 4 247 95
GenBank mRNA AF319614.1 - 1 - 2 113 95

View Full Product Details