Assay Design



Gene Transcripts

Gene Symbol
clh-3
Entrez Gene ID
174187
Gene Name
protein-coding
Gene Aliases
CELE_E04F6.11
Chromosome Location
Chr.II: 7217943 - 7225914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026889.5 NP_001022060.1 7 - 8 828 111
NM_001313600.3 NP_001300529.1 6 - 6 716 111
NM_001313601.3 NP_001300530.1 6 - 6 689 111
NM_001313602.3 NP_001300531.1 5 - 6 599 111
NM_001313603.2 NP_001300532.1 4 - 5 503 111
NM_001313604.3 NP_001300533.1 6 - 6 689 111
NM_001313605.3 NP_001300534.1 5 - 6 599 111
NM_001313606.3 NP_001300535.1 6 - 7 716 111
NR_132585.1 6 - 7 716 111
GenBank mRNA AF319614.1 - 4 - 5 582 111

View Full Product Details