Assay Design



Gene Transcripts

Gene Symbol
clh-3
Entrez Gene ID
174187
Gene Name
protein-coding
Gene Aliases
CELE_E04F6.11
Chromosome Location
Chr.II: 7217943 - 7225914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026889.5 NP_001022060.1 9 - 10 1377 123
NM_001313600.3 NP_001300529.1 8 - 8 1265 123
NM_001313601.3 NP_001300530.1 8 - 8 1238 123
NM_001313602.3 NP_001300531.1 7 - 8 1148 123
NR_132585.1 8 - 9 1265 123
GenBank mRNA AF319614.1 - 6 - 7 1131 123

View Full Product Details