Assay Design



Gene Transcripts

Gene Symbol
brf-1
Entrez Gene ID
174199
Gene Name
protein-coding
Gene Aliases
CELE_F45E12.2
Chromosome Location
Chr.II: 7305858 - 7308792 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322739.3 NP_001309557.1 7 - 8 2169 93
NM_001322740.4 NP_001309558.1 7 - 8 2157 93

View Full Product Details