Assay Design



Gene Transcripts

Gene Symbol
aka-1
Entrez Gene ID
174216
Gene Name
protein-coding
Gene Aliases
CELE_D1022.7
Chromosome Location
Chr.II: 7472606 - 7478539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026874.4 NP_001022045.1 15 - 16 3642 87
NM_001383888.1 16 - 17 3543 87
NM_063164.8 NP_495565.2 15 - 16 3654 87
GenBank mRNA AF045926.1 - 15 - 16 3653 87

View Full Product Details