Assay Design



Gene Transcripts

Gene Symbol
aka-1
Entrez Gene ID
174216
Gene Name
protein-coding
Gene Aliases
CELE_D1022.7
Chromosome Location
Chr.II: 7472606 - 7478539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026874.4 NP_001022045.1 5 - 6 1656 65
GenBank mRNA AF045926.1 - 5 - 6 1667 65

View Full Product Details