Assay Design



Gene Transcripts

Gene Symbol
aka-1
Entrez Gene ID
174216
Gene Name
protein-coding
Gene Aliases
CELE_D1022.7
Chromosome Location
Chr.II: 7472606 - 7478539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026874.4 NP_001022045.1 9 - 10 2813 102
NM_001383888.1 10 - 11 2700 102
NM_063164.8 NP_495565.2 9 - 10 2825 102
GenBank mRNA AF045926.1 - 9 - 10 2824 102

View Full Product Details