Assay Design



Gene Transcripts

Gene Symbol
nfi-1
Entrez Gene ID
174225
Gene Name
protein-coding
Gene Aliases
CELE_ZK1290.4
Chromosome Location
Chr.II: 7538180 - 7543007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027334.3 NP_001022505.1 8 - 9 1260 91
NM_001381498.1 NP_001367580.1 7 - 8 1208 91
NM_001381499.2 NP_001367579.1 7 - 8 1305 91

View Full Product Details