Assay Design



Gene Transcripts

Gene Symbol
tph-1
Entrez Gene ID
174227
Gene Name
protein-coding
Gene Aliases
CELE_ZK1290.2
Chromosome Location
Chr.II: 7549335 - 7551987 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373740.3 NP_001359899.1 7 - 7 1067 80
NM_063183.6 NP_495584.4 8 - 9 1069 80
GenBank mRNA AF135186.1 - 8 - 9 1089 80

View Full Product Details