Assay Design



Gene Transcripts

Gene Symbol
alfa-1
Entrez Gene ID
174236
Gene Name
protein-coding
Gene Aliases
CELE_F18A1.6
Chromosome Location
Chr.II: 7658325 - 7662522 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063202.8 NP_495603.1 3 - 4 445 124
NM_063203.6 NP_495604.1 2 - 3 450 124

View Full Product Details