Assay Design
Gene Transcripts
- Gene Symbol
- spdl-1
- Entrez Gene ID
-
174256
- Gene Name
- Spindly-like protein spdl-1
- Gene Aliases
-
CELE_C06A8.5
- Chromosome Location
-
Chr.II: 7781566 - 7783354 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063236.6 |
NP_495637.1 |
3 - 4 |
625 |
93 |
View Full Product Details