Assay Design



Gene Transcripts

Gene Symbol
cki-2
Entrez Gene ID
174261
Gene Name
protein-coding
Gene Aliases
CELE_T05A6.2
Chromosome Location
Chr.II: 7816499 - 7820281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027138.6 NP_001022309.1 4 - 5 623 64
NM_063241.7 NP_495642.3 4 - 5 621 64
GenBank mRNA AF179359.1 - 3 - 4 517 64

View Full Product Details