Assay Design
Gene Transcripts
- Gene Symbol
- lin-5
- Entrez Gene ID
-
174267
- Gene Name
- Spindle apparatus protein lin-5
- Gene Aliases
-
CELE_T09A5.10
- Chromosome Location
-
Chr.II: 7860659 - 7864153 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063253.8 |
NP_495654.1 |
2 - 3 |
759 |
80 |
GenBank mRNA |
AF240692.1 |
- |
2 - 3 |
797 |
80 |
View Full Product Details