Assay Design



Gene Transcripts

Gene Symbol
pgp-11
Entrez Gene ID
183971
Gene Name
P-GlycoProtein related
Gene Aliases
CELE_DH11.3
Chromosome Location
Chr.II: 7995580 - 8004257 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377827.2 NP_001364583.1 9 - 10 1763 84

View Full Product Details