Assay Design
Gene Transcripts
- Gene Symbol
- rpl-10
- Entrez Gene ID
-
174311
- Gene Name
- 60S ribosomal protein L10
- Gene Aliases
-
CELE_F10B5.1
- Chromosome Location
-
Chr.II: 8143881 - 8144683 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063306.8 |
NP_495707.1 |
2 - 3 |
433 |
85 |
View Full Product Details