Assay Design
Gene Transcripts
- Gene Symbol
- qua-1
- Entrez Gene ID
-
174319
- Gene Name
- Protein qua-1
- Gene Aliases
-
CELE_T05C12.10
- Chromosome Location
-
Chr.II: 8198803 - 8204005 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063324.4 |
NP_495725.2 |
9 - 10 |
3257 |
125 |
View Full Product Details