Assay Design
Gene Transcripts
- Gene Symbol
- rnh-2
- Entrez Gene ID
-
266861
- Gene Name
- Ribonuclease H2 subunit A
- Gene Aliases
-
CELE_T13H5.7
- Chromosome Location
-
Chr.II: 8517150 - 8518414 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063395.4 |
NP_495796.1 |
1 - 2 |
104 |
63 |
GenBank mRNA |
AF181619.1 |
- |
1 - 2 |
114 |
63 |
View Full Product Details