Assay Design



Gene Transcripts

Gene Symbol
glb-27
Entrez Gene ID
189091
Gene Name
protein-coding
Gene Aliases
CELE_W01C9.5
Chromosome Location
Chr.II: 8547337 - 8556266 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313555.3 NP_001300484.1 5 - 6 788 134
NM_001313670.4 NP_001300599.1 4 - 5 695 134
NM_063405.4 NP_495806.1 3 - 4 589 134

View Full Product Details