Assay Design
Gene Transcripts
- Gene Symbol
- stc-1
- Entrez Gene ID
-
174368
- Gene Name
- STCH (truncated HSP) family
- Gene Aliases
-
CELE_F54C9.2
- Chromosome Location
-
Chr.II: 8563709 - 8566139 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063407.6 |
NP_495808.2 |
7 - 8 |
1195 |
89 |
View Full Product Details