Assay Design
Gene Transcripts
- Gene Symbol
- cpf-1
- Entrez Gene ID
-
174378
- Gene Name
- WD_REPEATS_REGION domain-containing protein
- Gene Aliases
-
CELE_F28C6.3
- Chromosome Location
-
Chr.II: 8600449 - 8602479 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063421.6 |
NP_495822.1 |
1 - 2 |
119 |
78 |
View Full Product Details