Assay Design
Gene Transcripts
- Gene Symbol
- suf-1
- Entrez Gene ID
-
174380
- Gene Name
- Suf domain-containing protein
- Gene Aliases
-
CELE_F28C6.6
- Chromosome Location
-
Chr.II: 8603858 - 8606744 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063424.7 |
NP_495825.1 |
3 - 4 |
221 |
139 |
View Full Product Details