Assay Design



Gene Transcripts

Gene Symbol
cutl-12
Entrez Gene ID
188735
Gene Name
protein-coding
Gene Aliases
CELE_T22C8.7
Chromosome Location
Chr.II: 8630693 - 8634486 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322749.3 NP_001309578.1 3 - 3 364 80

View Full Product Details