Assay Design



Gene Transcripts

Gene Symbol
cutl-12
Entrez Gene ID
188735
Gene Name
protein-coding
Gene Aliases
CELE_T22C8.7
Chromosome Location
Chr.II: 8630693 - 8634486 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322749.3 NP_001309578.1 4 - 4 462 163
NM_001322750.3 NP_001309579.1 1 - 2 44 163

View Full Product Details