Assay Design



Gene Transcripts

Gene Symbol
prx-5
Entrez Gene ID
174392
Gene Name
protein-coding
Gene Aliases
CELE_C34C6.6
Chromosome Location
Chr.II: 8704822 - 8706768 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026848.5 NP_001022019.1 5 - 6 959 95
NM_001026849.3 NP_001022020.1 5 - 6 955 95

View Full Product Details