Assay Design
Gene Transcripts
- Gene Symbol
- nas-25
- Entrez Gene ID
-
174412
- Gene Name
- Zinc metalloproteinase nas-25
- Gene Aliases
-
CELE_F46C5.3
- Chromosome Location
-
Chr.II: 8826069 - 8828215 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063479.5 |
NP_495880.1 |
2 - 3 |
143 |
100 |
View Full Product Details