Assay Design



Gene Transcripts

Gene Symbol
set-17
Entrez Gene ID
174425
Gene Name
protein-coding
Gene Aliases
CELE_T21B10.5
Chromosome Location
Chr.II: 8939054 - 8940488 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330955.2 NP_001317778.1 2 - 3 204 79
NM_001381544.1 NP_001368599.1 2 - 3 168 79

View Full Product Details