Assay Design
Gene Transcripts
- Gene Symbol
- gcs-1
- Entrez Gene ID
-
174438
- Gene Name
- Glutamate--cysteine ligase
- Gene Aliases
-
CELE_F37B12.2
- Chromosome Location
-
Chr.II: 9031854 - 9038301 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063526.8 |
NP_495927.2 |
5 - 6 |
1701 |
87 |
View Full Product Details