Assay Design
Gene Transcripts
- Gene Symbol
- sec-5
- Entrez Gene ID
-
174459
- Gene Name
- Exocyst complex component 2
- Gene Aliases
-
CELE_T23G7.4
- Chromosome Location
-
Chr.II: 9173161 - 9177152 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063557.6 |
NP_495958.1 |
11 - 12 |
2548 |
102 |
View Full Product Details