Assay Design
Gene Transcripts
- Gene Symbol
- gst-13
- Entrez Gene ID
-
188915
- Gene Name
- Glutathione S-Transferase
- Gene Aliases
-
CELE_T26C5.1
- Chromosome Location
-
Chr.II: 9231457 - 9232344 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063566.7 |
NP_495967.1 |
2 - 3 |
633 |
111 |
View Full Product Details