Assay Design
Gene Transcripts
- Gene Symbol
- kin-16
- Entrez Gene ID
-
174499
- Gene Name
- Receptor-like tyrosine-protein kinase kin-16
- Gene Aliases
-
CELE_M176.7
- Chromosome Location
-
Chr.II: 9434017 - 9437207 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063616.6 |
NP_496017.1 |
7 - 8 |
1324 |
69 |
View Full Product Details