Assay Design



Gene Transcripts

Gene Symbol
gcy-3
Entrez Gene ID
191641
Gene Name
protein-coding
Gene Aliases
CELE_R134.1
Chromosome Location
Chr.II: 9502757 - 9507281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377837.1 NP_001364551.1 5 - 6 865 128
NM_063636.1 NP_496037.1 5 - 6 865 128

View Full Product Details