Assay Design
Gene Transcripts
- Gene Symbol
- mex-6
- Entrez Gene ID
-
174504
- Gene Name
- Zinc finger protein mex-6
- Gene Aliases
-
CELE_AH6.5
- Chromosome Location
-
Chr.II: 9524174 - 9526390 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063642.6 |
NP_496043.1 |
1 - 2 |
139 |
61 |
View Full Product Details