Assay Design
Gene Transcripts
- Gene Symbol
- ugt-58
- Entrez Gene ID
-
174507
- Gene Name
- Putative UDP-glucuronosyltransferase ugt-58
- Gene Aliases
-
CELE_F35H8.6
- Chromosome Location
-
Chr.II: 9584380 - 9586784 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063658.8 |
NP_496059.2 |
1 - 2 |
181 |
70 |
View Full Product Details