Assay Design
Gene Transcripts
- Gene Symbol
- cyp-13A6
- Entrez Gene ID
-
188357
- Gene Name
- Putative cytochrome P450 CYP13A6
- Gene Aliases
-
CELE_T10B9.3
- Chromosome Location
-
Chr.II: 9800758 - 9802821 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063712.4 |
NP_496113.1 |
1 - 2 |
276 |
83 |
View Full Product Details