Assay Design
Gene Transcripts
- Gene Symbol
- old-2
- Entrez Gene ID
-
191738
- Gene Name
- Protein kinase domain-containing protein
- Gene Aliases
-
CELE_ZK938.5
- Chromosome Location
-
Chr.II: 9847483 - 9849597 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063722.1 |
NP_496123.1 |
5 - 6 |
936 |
102 |
View Full Product Details