Assay Design



Gene Transcripts

Gene Symbol
egl-43
Entrez Gene ID
174552
Gene Name
protein-coding
Gene Aliases
CELE_R53.3
Chromosome Location
Chr.II: 9955185 - 9962995 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027117.6 NP_001022288.1 9 - 10 1492 99
NM_001373787.1 NP_001359539.1 8 - 9 1373 99

View Full Product Details