Assay Design
Gene Transcripts
- Gene Symbol
- srb-6
- Entrez Gene ID
-
191788
- Gene Name
- Serpentine receptor class beta-6
- Gene Aliases
-
CELE_R05H5.6
- Chromosome Location
-
Chr.II: 10194905 - 10196835 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063798.1 |
NP_496199.1 |
5 - 6 |
642 |
138 |
View Full Product Details