Assay Design
Gene Transcripts
- Gene Symbol
- clec-60
- Entrez Gene ID
-
191384
- Gene Name
- VWFA domain-containing protein
- Gene Aliases
-
CELE_ZK666.6
- Chromosome Location
-
Chr.II: 10480043 - 10483138 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063858.3 |
NP_496259.1 |
4 - 5 |
446 |
92 |
View Full Product Details