Assay Design
Gene Transcripts
- Gene Symbol
- ect-2
- Entrez Gene ID
-
174658
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T19E10.1
- Chromosome Location
-
Chr.II: 10778387 - 10783962 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063917.6 |
NP_496318.1 |
12 - 13 |
2410 |
66 |
View Full Product Details