Assay Design



Gene Transcripts

Gene Symbol
ect-2
Entrez Gene ID
174658
Gene Name
protein-coding
Gene Aliases
CELE_T19E10.1
Chromosome Location
Chr.II: 10778387 - 10783962 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063917.6 NP_496318.1 3 - 4 481 66
NM_063918.3 NP_496319.1 4 - 5 506 66

View Full Product Details